4-Hydroxyzimtsäure 98%
Bottle
Artikelnr. APOSOR21362-2.5KG
New Product
Anbieter: Apollo Scientific
4-Hydroxyzimtsäure 98%
Artikelnr. APOSOR21362-2.5KG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 7400-08-0
Springe zu:
Product Details & Documents
Synonyme: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
- CAS Nummer:7400-08-0
- Formel:C₉H₈O₃
- MDL-Nummer:MFCD00004399
- Molekulargewicht:164.16 g/mol
- Siedepunkt:346 °C
- Schmelzpunkt:212…214 °C
- Lagertemperatur:Raumtemperatur
Spezifikationen
Spezifikationen
- Packung:Bottle
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



