DIPSO ≥98%
Bottle
Artikelnr. APOSBIB1158-25G
New Product
Anbieter: Apollo Scientific
Springe zu:
Product Details & Documents
Biological buffer - useful range 7.0 - 8.2.
US Patent No. 4,169,950 (To Research Organics)
Synonyme: 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
- CAS Nummer:68399-80-4
- Formel:C₇H₁₇NO₆S
- MDL-Nummer:MFCD00038353
- Molekulargewicht:261.29 g/mol
- Schmelzpunkt:189…192 °C
- Lagertemperatur:Raumtemperatur
Spezifikationen
Spezifikationen
- Packung:Bottle
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



