Express Eingabe
Austria
AuftragsverfolgungKontakt
 

1,2-Dibenzyldiselane 98%

Bottle
Catalog # APOSOR89730-5G
New Product
Supplier:   Apollo Scientific
1,2-Dibenzyldiselane 98%
Catalog # APOSOR89730-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  1482-82-2
1,2-Dibenzyldiselane 98%

Bottle

CAS Number:  1482-82-2
Price
26,10 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:1482-82-2
  • Formula:C₁₄H₁₄Se₂
  • MDL Number:MFCD00004767
  • Molecular Weight:340,19 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications