Express Eingabe
Austria
AuftragsverfolgungKontakt
1,2-Dibenzyldiselane 98%

1,2-Dibenzyldiselane 98%

Bottle
Catalog # APOSOR89730-1G
New Product
Supplier:   Apollo Scientific
1,2-Dibenzyldiselane 98%
Catalog # APOSOR89730-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1482-82-2
1,2-Dibenzyldiselane 98%

Bottle

CAS Number:  1482-82-2
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:1482-82-2
  • Formula:C₁₄H₁₄Se₂
  • MDL Number:MFCD00004767
  • Molecular Weight:340,19 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.