1,2-Dibenzyldiselane 98%
Bottle
Artikelnr. APOSOR89730-500G
New Product
Anbieter: Apollo Scientific
1,2-Dibenzyldiselane 98%
Artikelnr. APOSOR89730-500G
Anbieter: Apollo Scientific
New Product
CAS Nummer: 1482-82-2
Springe zu:
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Nummer:1482-82-2
- Formel:C₁₄H₁₄Se₂
- MDL-Nummer:MFCD00004767
- Molekulargewicht:340,19 g/mol
Spezifikationen
Spezifikationen
- Packung:Bottle
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



