A-196 ≥98%
Bottle
Artikelnr. APOSBUP05604-50MG
New Product
Anbieter: Apollo Scientific
A-196 ≥98%
Artikelnr. APOSBUP05604-50MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 1982372-88-2
Springe zu:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyme: A-196
- CAS Nummer:1982372-88-2
- Formel:C₁₈H₁₆Cl₂N₄
- Molekulargewicht:359,25 g/mol
Spezifikationen
Spezifikationen
- Packung:Bottle
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



