Express Eingabe
Austria
AuftragsverfolgungKontakt
Pifithrin-α hydrobromide ≥95%

Pifithrin-α hydrobromide ≥95%

Bottle
Catalog # APOSBUP05923-100MG
New Product
Supplier:   Apollo Scientific
Pifithrin-α hydrobromide ≥95%
Catalog # APOSBUP05923-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  63208-82-2
Pifithrin-α hydrobromide ≥95%

Bottle

CAS Number:  63208-82-2
Price
558,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
  • CAS Number:63208-82-2
  • Formula:C₁₆H₁₈N₂OS·HBr
  • Molecular Weight:367,3 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.