Pifithrin-α hydrobromide ≥95%
Bottle
Artikelnr. APOSBUP05923-50MG
New Product
Anbieter: Apollo Scientific
Pifithrin-α hydrobromide ≥95%
Artikelnr. APOSBUP05923-50MG
Anbieter: Apollo Scientific
New Product
CAS Nummer: 63208-82-2
Springe zu:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyme: Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
- CAS Nummer:63208-82-2
- Formel:C₁₆H₁₈N₂OS·HBr
- Molekulargewicht:367,3 g/mol
Spezifikationen
Spezifikationen
- Packung:Bottle
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



