Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Pifithrin-α hydrobromide ≥95%

Bottle
Catalog # APOSBUP05923-25MG
New Product
Supplier:   Apollo Scientific
Pifithrin-α hydrobromide ≥95%
Catalog # APOSBUP05923-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  63208-82-2
Pifithrin-α hydrobromide ≥95%

Bottle

CAS Number:  63208-82-2
Price
231,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
  • CAS Number:63208-82-2
  • Formula:C₁₆H₁₈N₂OS·HBr
  • Molecular Weight:367,3 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications