D-Mannose ≥95%
Bottle
Catalog # APOSBUP16831-5G
New Product
Supplier: Apollo Scientific
D-Mannose ≥95%
Catalog # APOSBUP16831-5G
Supplier: Apollo Scientific
New Product
CAS Number: 3458-28-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- Molecular Weight:180,16 g/mol
Specifications
Specifications
- Packung:Bottle