AuftragsverfolgungKontakt
D-Mannose ≥95%
Bottle
Catalog # APOSBUP16831-1G
New Product
Supplier:   Apollo Scientific
D-Mannose ≥95%
Catalog # APOSBUP16831-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  3458-28-4
D-Mannose ≥95%
Bottle
CAS Number:  3458-28-4
Price
170,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • Molecular Weight:180,16 g/mol
Specifications
Specifications
  • Packung:
    Bottle
Specifications