Xanthydrol
Bottle
Artikelnr. APOSBIX1001-500G
New Product
Anbieter: Apollo Scientific
Springe zu:
Product Details & Documents
Synonyme: 9-Hydroxyxanthene, 9-Xanthenol, 9H-Xanthen-9-ol
- CAS Nummer:90-46-0
- Formel:C₁₃H₁₀O₂
- MDL-Nummer:MFCD00005057
- Molekulargewicht:198.22 g/mol
- Schmelzpunkt:124…127 °C
- Lagertemperatur:Kühlschrank
Spezifikationen
Spezifikationen
- Packung:Bottle
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



