Naringin
Naringin
EHERC15495000
- SMILES: C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)C[C@H](Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O
- Inchi: InChI=1S/C27H32O14/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11/h2-7,10,16,18,20-30,32-36H,8-9H2,1H3/t10-,16-,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1
- IUPAC: (2S)-7-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
- Bezeichnung:Naringin
- Cat. No.:EHERC15495000
- CAS Nr.:10236-47-2
- Molekulargewicht:580,53
- Lagerbedingungen:4 °C
- Synonym(s):4H-1-Benzopyran-4-one, 7-[[2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,3-dihydro-5-hydroxy-2-(4-hydroxyphenyl)-, (2S)-;4H-1-Benzopyran-4-one, 7-[[2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,3-dihydro-5-hydroxy-2-(4-hydroxyphenyl)-, (S)-;Naringin (6CI,7CI,8CI);(2S)-Naringin;7-[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyloxy]-2,3-dihydro-4',5,7-trihydroxyflavone;Aurantiin;NSC 5548;Naringenin 7-O-neohesperidoside;Naringenin 7-neohesperidoside;Naringenin 7-β-neohesperidoside;Naringoside
- Shipping conditions:RT
- Formel:C₂₇H₃₂O₁₄



