Imibenconazole
Imibenconazole
Artikelnr. EHERC14283600
Anbieter: EHRENSTORFER
Springe zu:
Product Details & Documents
- SMILES: Clc1ccc(CS\C(=N\c2ccc(Cl)cc2Cl)\Cn3cncn3)cc1
- Inchi: InChI=1S/C17H13Cl3N4S/c18-13-3-1-12(2-4-13)9-25-17(8-24-11-21-10-22-24)23-16-6-5-14(19)7-15(16)20/h1-7,10-11H,8-9H2/b23-17+
- IUPAC: (4-chlorophenyl)methyl (1E)-N-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-yl)ethanimidothioate
- Packing group: III
Spezifikationen
Spezifikationen
- Bezeichnung:Imibenconazole
- UN No.:3077
- Cat. No.:EHERC14283600
- CAS Nr.:86598-92-7
- GHS-Signal:Achtung
- Molekulargewicht:411,74
- Beschreibung Gefahrenklasse:GHS07, GHS09
- Lagerbedingungen:–18 °C
- Shipping conditions:RT
- Formel:C₁₇H₁₃Cl₃N₄S
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



