Acetophenone
Acetophenone
Artikelnr. EHERC10022000
Anbieter: EHRENSTORFER
Springe zu:
Product Details & Documents
- SMILES: CC(=O)c1ccccc1
- Inchi: InChI=1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3
- IUPAC: 1-phenylethanone
Spezifikationen
Spezifikationen
- Bezeichnung:Acetophenone
- Cat. No.:EHERC10022000
- CAS Nr.:98-86-2
- GHS-Signal:Achtung
- Molekulargewicht:120,15
- Beschreibung Gefahrenklasse:GHS07
- Lagerbedingungen:20 °C
- Synonym(s):Acetophenone;Acetophenone (8CI);1-Phenylethanone;1-Feniletanone;1-Phenyl-1-ethanone;Acetophenon;Acetylbenzene;Benzoylmethane;Hypnon;Hypnone;Methyl phenyl ketone;NSC 7635;NSC 98542;Phenyl methyl ketone
- Shipping conditions:RT
- Formel:C₈H₈O
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



