Sulfanitran
Sulfanitran
Artikelnr. EHERC17000050
Anbieter: EHRENSTORFER
Springe zu:
Product Details & Documents
- SMILES: CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2ccc(cc2)[N+](=O)[O-] Inchi: InChI=1S/C14H13N3O5S/c1-10(18)15-11-4-8-14(9-5-11)23(21,22)16-12-2-6-13(7-3-12)17(19)20/h2-9,16H,1H3,(H,15,18) IUPAC: N-[4-[(4-nitrophenyl)sulfamoyl]phenyl]acetamide
Spezifikationen
Spezifikationen
- Bezeichnung:Sulfanitran
- Cat. No.:EHERC17000050
- CAS Nr.:122-16-7
- GHS-Signal:Achtung
- Molekulargewicht:335,34
- Beschreibung Gefahrenklasse:GHS07
- Lagerbedingungen:20 ℃
- Synonym(s):Acetamide, N-[4-[[(4-nitrophenyl)amino]sulfonyl]phenyl]- (9CI, ACI); Acetanilide, 4'-[(p-nitrophenyl)sulfamoyl]- (6CI, 7CI, 8CI); N-[4-[[(4-Nitrophenyl)amino]sulfonyl]phenyl]acetamide (ACI); 4'-[(p-Nitrophenyl)sulfamoyl]acetanilide; N4-Acetyl-4'-nitrosulfanilanilide; N4-Acetyl-N1-(4-nitrophenyl)sulfanilamide; N4-Acetyl-N1-(p-nitrophenyl)sulfanilamide; NSC 217299; NSC 77120; Sulfanitran
- Shipping conditions:RT
- Formel:C₁₄H₁₃N₃O₅S
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



