Pentabromophenol
Pentabromophenol
Artikelnr. EHERC15939300
Anbieter: EHRENSTORFER
Springe zu:
Product Details & Documents
- SMILES: Oc1c(Br)c(Br)c(Br)c(Br)c1Br Inchi: InChI=1S/C6HBr5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H IUPAC: 2,3,4,5,6-pentabromophenol
- Packing group: III
Spezifikationen
Spezifikationen
- Bezeichnung:Pentabromophenol
- UN No.:2811
- Cat. No.:EHERC15939300
- CAS Nr.:608-71-9
- GHS-Signal:Gefahr
- Molekulargewicht:488,59
- Beschreibung Gefahrenklasse:GHS06, GHS07, GHS09
- Lagerbedingungen:20 ℃
- Synonym(s):Phenol, 2,3,4,5,6-pentabromo-; Phenol, pentabromo- (6CI,7CI,8CI,9CI); 2,3,4,5,6-Pentabromophenol; Bromophenasic acid; Flammex 5BP; NSC 5717; Pentabromophenol; Perbromophenol
- Shipping conditions:RT
- Formel:C₆HBr₅O
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



