Ethynylbenzene
Ethynylbenzene
Artikelnr. EHERC13356450
Anbieter: EHRENSTORFER
Springe zu:
Product Details & Documents
- SMILES: C#Cc1ccccc1 Inchi: InChI=1S/C8H6/c1-2-8-6-4-3-5-7-8/h1,3-7H IUPAC: ethynylbenzene
- Packing group: III
Spezifikationen
Spezifikationen
- Bezeichnung:Ethynylbenzene
- UN No.:3295
- Cat. No.:EHERC13356450
- CAS Nr.:536-74-3
- GHS-Signal:Gefahr
- Molekulargewicht:102,13
- Beschreibung Gefahrenklasse:GHS02, GHS07, GHS08
- Lagerbedingungen:20 ℃
- Synonym(s):Ethynylbenzene; 1-Ethynylbenzene; 1-Phenylethyne; Ethyne, phenyl-; NSC 4957; Phenylacetylene; Phenylethyne
- Shipping conditions:20 ℃
- Formel:C₈H₆
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



