2,4,5-Trichlorophenol
2,4,5-Trichlorophenol
Catalog # EHERC17774500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Oc1cc(Cl)c(Cl)cc1Cl Inchi: InChI=1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H IUPAC: 2,4,5-trichlorophenol
- Packing group: III
Specifications
Specifications
- Bezeichnung:2,4,5-Trichlorophenol
- UN No.:2020
- Cat. No.:EHERC17774500
- CAS Nr.:95-95-4
- GHS-Signal:Achtung
- Molekulargewicht:197,45
- Beschreibung Gefahrenklasse:GHS07, GHS09
- Lagerbedingungen:20 ℃
- Synonym(s):Phenol, 2,4,5-trichloro-; 2,4,5-Trichlorophenol; 2.4,5-Trichlorophenic acid; Dowicide 2; NSC 2266; Preventol I; TCP
- Shipping conditions:RT
- Formel:C₆H₃Cl₃O



