Transfluthrin
Transfluthrin
Catalog # EHERC17606000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1(C)[C@H](C=C(Cl)Cl)[C@H]1C(=O)OCc2c(F)c(F)cc(F)c2F
- Inchi: InChI=1S/C15H12Cl2F4O2/c1-15(2)7(3-10(16)17)11(15)14(22)23-5-6-12(20)8(18)4-9(19)13(6)21/h3-4,7,11H,5H2,1-2H3/t7-,11+/m1/s1
- IUPAC: (2,3,5,6-tetrafluorophenyl)methyl (1R,3S)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
- Packing group: III
Specifications
Specifications
- Bezeichnung:Transfluthrin
- UN No.:3349
- Cat. No.:EHERC17606000
- CAS Nr.:118712-89-3
- GHS-Signal:Gefahr
- Molekulargewicht:371,15
- Beschreibung Gefahrenklasse:GHS06, GHS07, GHS09
- Lagerbedingungen:20 °C
- Shipping conditions:RT
- Formel:C₁₅H₁₂Cl₂F₄O₂



