Methyl pentachlorophenylsulfide
Methyl pentachlorophenylsulfide
Artikelnr. EHERC15120000
Anbieter: EHRENSTORFER
Springe zu:
Product Details & Documents
- SMILES: CSc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl
- Inchi: InChI=1S/C7H3Cl5S/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h1H3
- IUPAC: 1,2,3,4,5-pentachloro-6-methylsulfanylbenzene
Spezifikationen
Spezifikationen
- Bezeichnung:Methyl pentachlorophenylsulfide
- Cat. No.:EHERC15120000
- CAS Nr.:1825-19-0
- GHS-Signal:Achtung
- Molekulargewicht:296,43
- Beschreibung Gefahrenklasse:GHS07
- Lagerbedingungen:20 °C
- Synonym(s):Benzene;pentachloro(methylthio)- (9CI);Sulfide;methyl pentachlorophenyl (6CI,7CI,8CI);1,2,3,4,5-Pentachloro-6-(methylthio)benzene;Methyl pentachlorophenyl sulfide;Methylthiopentachlorobenzene;PCTA;Pentachloro(methylthio)benzene;Pentachlorophenyl methyl sulfide;Pentachlorothioanisole
- Shipping conditions:Room temperature
- Formel:C₇H₃Cl₅S
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



