DMST
DMST
- Bezeichnung:DMST
- Cat. No.:EHERC13040000
- CAS Nr.:66840-71-9
- GHS-Signal:Achtung
- Molekulargewicht:214,28
- Beschreibung Gefahrenklasse:GHS09
- Lagerbedingungen:20 °C
- Synonym(s):N,N-dimethyl-N'-(4-methylphenyl)sulfamide;Sulfamide;N,N-dimethyl-N'-p-tolyl- (6CI);N,N-Dimethyl-N'-(4-methylphenyl)sulfamide;4-Dimethylaminosulfotoluidide;DMST;N,N-dimethyl-N'-p-tolylsulfamide
- Shipping conditions:RT
- Formel:C₉H₁₄N₂O₂S
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



