3,3'-Dimethoxybenzidine 100 µg/ml in Acetonitrile
3,3'-Dimethoxybenzidine 100 µg/ml in Acetonitrile
Artikelnr. EHERA12721000AL
Anbieter: EHRENSTORFER
3,3'-Dimethoxybenzidine 100 µg/ml in Acetonitrile
Artikelnr. EHERA12721000AL
Anbieter: EHRENSTORFER
CAS Nummer:
Springe zu:
Product Details & Documents
- SMILES: COc1cc(ccc1N)c2ccc(N)c(OC)c2
- Inchi: InChI=1S/C14H16N2O2/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2/h3-8H,15-16H2,1-2H3
- IUPAC: 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
- Packing group: II
Spezifikationen
Spezifikationen
- Bezeichnung:3,3'-Dimethoxybenzidine 100 µg/ml in Acetonitrile
- UN No.:1648M
- Cat. No.:EHERA12721000AL
- CAS Nr.:119-90-4
- GHS-Signal:Gefahr
- Molekulargewicht:244,29
- Beschreibung Gefahrenklasse:GHS02, GHS07
- Lagerbedingungen:–18 °C
- Synonym(s):Benzidine;3,3'-dimethoxy- (8CI);Fast Blue B Base (6CI);3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diamine;3,3'-Dimethoxy-4,4'-diaminobiphenyl;3,3'-Dimethoxy-4,4'-diaminodiphenyl;4,4'-Bi-o-anisidine;4,4'-Diamino-3,3'-dimethoxy-1,1'-biphenyl;Amacel Developed Navy SD;Azogene Fast Blue B;Blue BN Base;Blue Base Irga B;Blue Base NB;C.I. Disperse Black 6;Cellitazol B;Cibacete Diazo Navy Blue 2B;Diacel Navy DC;Dianisidine;Fast Blue Base B;Fast Blue DSC Base;Hiltonil Fast Blue B Base;Kayaku Blue B Base;Lake Blue B Base;Mitsui Blue B Base;NSC 3168;Naphthanil Blue B Base;Setacyl Diazo Navy R;o-Dianisidine
- Shipping conditions:RT
- Formel:C₁₄H₁₆N₂O₂
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



