Levonorgestrel
Levonorgestrel
EHERC14629760
- SMILES: CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
- Inchi: InChI=1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1
- IUPAC: (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
- Bezeichnung:Levonorgestrel
- Cat. No.:EHERC14629760
- CAS Nr.:797-63-7
- GHS-Signal:Gefahr
- Molekulargewicht:312,45
- Beschreibung Gefahrenklasse:GHS08
- Lagerbedingungen:4 °C
- Synonym(s):Levonorgestrel; (-)-Norgestrel; 18-Methylnorethindrone; 19-Nortestosterone, 17-ethynyl-18-methyl-; D-(-)-Norgestrel; D-Norgestrel; Dexnorgestrel; Femilis; Femilis Slim; Follistrel; Jadelle; Levonova; Lng-IUS; Microlut; Microluton; Microval; Mirena; Norgeston; Norgestrelem; Norlevo; Norplant; Norplant 2; Norplant II; Postinor 2; Wy 5104; (17alpha)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one; 13-Ethyl-17-hydroxy-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one; 13-Ethyl-17-ethynyl-17beta-hydroxy-4-gonen-3-one; 13-Ethyl-17alpha-ethynyl-17-hydroxygon-4-en-3-one; 13-Ethyl-17alpha-ethynylgon-4-en-17beta-ol-3-one; 13beta-Ethyl-17alpha-ethynyl-17beta-hydroxygon-4-en-3-one; 17alpha-Ethynyl-13-ethyl-19-nortestosterone; 17alpha-Ethynyl-13beta-ethyl-3-oxo-4-estren-17beta-ol; 17alpha-Ethynyl-17-hydroxy-18-methylestr-4-en-3-one; 17alpha-Ethynyl-18-homo-19-nortestosterone; 17alpha-Ethynyl-18-methyl-19-nortestosterone; 17beta-Hydroxy-18-methyl-19-nor-17alpha-pregn-4-en-20-yn-3-one; 18-Methyl-17alpha-ethynyl-19-nortestosterone; d-(17alpha)-17-Hydroxy-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one; Norgestimate Impurity B
- Shipping conditions:20 °C
- Formel:C₂₁H₂₈O₂



