Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Ipronidazole-hydroxy

Ipronidazole-hydroxy
  EHERC14370720
Ipronidazole-hydroxy
  EHERC14370720
 :  
Ipronidazole-hydroxy
Ipronidazole-hydroxy

Ipronidazole-hydroxy

 :  
 
262,00 €
   
1 * 10 mg
 
  • SMILES: Cn1c(cnc1C(C)(C)O)[N+](=O)[O-]
  • Inchi: InChI=1S/C7H11N3O3/c1-7(2,11)6-8-4-5(9(6)3)10(12)13/h4,11H,1-3H3
  • IUPAC: 2-(1-methyl-5-nitroimidazol-2-yl)propan-2-ol
 

 

  • Bezeichnung:
    Ipronidazole-hydroxy
  • Cat. No.:
    EHERC14370720
  • CAS Nr.:
    35175-14-5
  • GHS-Signal:
    Achtung
  • Molekulargewicht:
    185,18
  • Beschreibung Gefahrenklasse:
    GHS07
  • Lagerbedingungen:
    20 °C
  • Synonym(s):
    α,α,1-Trimethyl-5-nitro-1H-imidazole-2-methanol; 1,α,α-Trimethyl-5-nitroimidiazole-2-methanol; 1-Methyl-2-(2'-hydroxyisopropyl)-5-nitroimidazole; 2-(1-Hydroxy-1-methylethyl)-1-methyl-5-nitroimidazole; Hydroxyipronidazole; Ipronidazole-OH; L 12407
  • Shipping conditions:
    RT
  • Formel:
    C₇H₁₁N₃O₃

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.