Digoxin
Digoxin
EHERC12633850
- SMILES: O[C@@]12[C@@](C)([C@H](CC1)C3=CC(=O)OC3)[C@H](O)C[C@]4([C@]2(CC[C@]5([C@]4(C)CC[C@H](O[C@H]6C[C@H](O)[C@H](O[C@H]7C[C@H](O)[C@H](O[C@H]8C[C@H](O)[C@H](O)[C@@H](C)O8)[C@@H](C)O7)[C@@H](C)O6)C5)[H])[H])[H]
- Inchi: InChI=1S/C41H64O14/c1-19-36(47)28(42)15-34(50-19)54-38-21(3)52-35(17-30(38)44)55-37-20(2)51-33(16-29(37)43)53-24-8-10-39(4)23(13-24)6-7-26-27(39)14-31(45)40(5)25(9-11-41(26,40)48)22-12-32(46)49-18-22/h12,19-21,23-31,33-38,42-45,47-48H,6-11,13-18H2,1-5H3/t19-,20-,21-,23-,24+,25-,26-,27+,28+,29+,30+,31-,33+,34+,35+,36-,37-,38-,39+,40+,41+/m1/s1
- IUPAC: 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-3-[(2R,4S,5S,6R)-5-[(2S,4S,5S,6R)-5-[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
- Packing group: II
- Bezeichnung:Digoxin
- UN No.:2811
- Cat. No.:EHERC12633850
- CAS Nr.:20830-75-5
- GHS-Signal:Gefahr
- Molekulargewicht:780,94
- Beschreibung Gefahrenklasse:GHS06, GHS08
- Lagerbedingungen:20 °C
- Synonym(s):Digoxin (8CI); Cardigox; Cardiogoxin; Cardioxin; Cardixin; Cardoxin; Chloroformic digitalin; Coragoxine; Cordioxil; Davoxin; Digacin; Digicor; Digitek; Digomal; Digon; Digosin; Digoxin Nativelle; Digoxine; Dilanacin; Dixina; Dokim; Dynamos; Eudigox; Fargoxin; Grexin; Homolle's digitalin; Lanacordin; Lanacrist; Lanicor; Lanikor; Lanocardin; Lanorale; Lanoxicaps; Lanoxin; Lanoxin PG; Lenoxicaps; Lenoxin; Longdigox; Mapluxin; NSC 95100; Natigoxin; NeoDioxanin; Novodigal-Amp.; Purgoxin; Rougoxin; Stillacor; Toloxin; Vanoxin; Card-20(22)-enolide, 3-[(O-2,6-dideoxy-beta-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-beta-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-beta-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxy-, (3beta,5beta,12beta)-; Digitoxin, 12beta-hydroxy- (6CI); (3beta,5beta,12beta)-3-[(O-2,6-Dideoxy-beta-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-beta-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-beta-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxycard-20(22)-enolide; 12beta-Hydroxydigitoxin; Digitoxin, 12β-hydroxy- (6CI); (3β,5β,12β)-3-[(O-2,6-Dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxycard-20(22)-enolide; 12β-Hydroxydigitoxin; 4-[(3S,5R,8R,9S,10S,12R,13S,14S)-3-[(2S,4S,5R,6R)-5-[(2S,4S,5R,6R)-5-[ (2S,4S,5R,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[ a]phenanthren-17-yl]-5H-furan-2-one
- Shipping conditions:RT
- Formel:C₄₁H₆₄O₁₄
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



