Hydroxyurea ≥98%
New Product
Anbieter: Apollo Scientific
Springe zu:
Über diesen Artikel
Usage: Biochemical/life science applications.
Synonyme: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Nummer:127-07-1
- Formel:CH₄N₂O₂
- Molekulargewicht:76,05 g/mol
2 Optionen unten verfügbar
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyme: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Nummer:127-07-1
- Formel:CH₄N₂O₂
- Molekulargewicht:76,05 g/mol
Product Family Options
Produktinformationen