Pifithrin-α hydrobromide ≥95%
New Product
Anbieter: Apollo Scientific
Springe zu:
Über diesen Artikel
Usage: Biochemical/life science applications.
Synonyme: Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
- CAS Nummer:63208-82-2
- Formel:C₁₆H₁₈N₂OS·HBr
- Molekulargewicht:367,3 g/mol
3 Optionen unten verfügbar
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyme: Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
- CAS Nummer:63208-82-2
- Formel:C₁₆H₁₈N₂OS·HBr
- Molekulargewicht:367,3 g/mol
Product Family Options
Produktinformationen
- PackungVerfügbarkeitPreis
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
- Spezifikationen:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



