Microbore Extension Set
Microbore extension set with rotating Luer-lock, PVC, 153 cm
Artikelnr. BDAMPA-150-G
New Product
Anbieter: BD Medical
Springe zu:
Product Details & Documents
BD provides a range of extension sets in small bore and standard bore tubing sizes with varying priming volumes depending on the tubing size and length.
Spezifikationen
- Bezeichnung:Microbore extension set with rotating Luer-lock
- Länge:153 cm
- Material:PVC