Combination Breathing Filter
Atemfilter, 345 g, 108Ø×93H mm
Artikelnr. BART202419
New Product
Anbieter: BARTELS RIEGER
Springe zu:
Product Details & Documents
Screw filter with round thread connection according to DIN EN 148-1, for protection against organic and inorganic gases and vapors, hydrogen chloride, sulfur dioxide, and ammonia, with simultaneous occurrence of particles of less toxic substances.
Individually sealed in PE bags and packed in outer carton.
Zertifizierung: DIN EN 133, DIN EN 148-1, DIN EN 149, DIN EN 14387.
Spezifikationen
- Bezeichnung:Atemfilter, 345 g, 108Ø×93H mm