AuftragsverfolgungKontakt
b.safe Trichter Control Set | Avantor
b.safe Trichter Control Set
Trichter Control Set, S60
Artikelnr. BOHLR9970-07
New Product
Anbieter:   Bohlender
b.safe Trichter Control Set
Artikelnr. BOHLR9970-07
Anbieter:  Bohlender
New Product
b.safe Trichter Control Set
b.safe Trichter Control Set
Trichter Control Set, S60
Preis
280,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Set consisting of b.safe Funnel 130 with integrated level indicator and matching 10-liter canister with thread S60 for the disposal of liquid HPLC waste.
All materials used are made of static dissipative PE-EX, with professional earthing of the system, static charging is avoided. The level indicator integrated in the funnel consists of a float with a red indicator pin. As soon as the indicator pin is visible above the top edge of the sieve, the waste container has to be exchanged to avoid spillover.
The sieve, which can be removed for cleaning, restrains magnetic stirring bars, for example, or other solids while the hinged lid prevents harmful vapours from escaping from the canister. Thanks to the inclined cap, the funnel can be positioned horizontally on the canister with inclined filling neck.
Delivery information: Supplied with 1×b.safe PE-HD-EX Funnel 130, with level control, 1×b.safe PE-HD-EX Canister S60 inclined, volume 10 liters.
Spezifikationen
  • Gewinde:
    S60
  • Bezeichnung:
    Trichter Control Set