Express Eingabe
Austria
AuftragsverfolgungKontakt
PF-9366 ≥98%
  ABCAAB286964-5
Marketsource
 :   Abcam
PF-9366 ≥98%
  ABCAAB286964-5
 :  Abcam
Marketsource
 :  
PF-9366 ≥98%
 :  
205,00 €
1 * 5 mg
An allosteric inhibitor of human methionine adenosyltransferase 2A (Mat2A), the extrahepatic isoform (IC₅₀ = 420 nM). PF-9366 inhibits cellular S-Adenosyl-L-methionine (SAM) production with an IC₅₀ of 1,2 μM in H520 lung carcinoma cells.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  2-(7-Chloro-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-1-yl)-N,N-dimethylethan-1-amine
  •  :350,8 g/mol