Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Pifithrin-α hydrobromide ≥95% p53 Inhibitor

Marketsource
Anbieter:   Abcam
Pifithrin-α hydrobromide ≥95% p53 Inhibitor
Anbieter:  Abcam
Marketsource
Zertifikate

2 Optionen unten verfügbar

Product Details & Documents

p53 Inhibitor. Specifically blocks tumour supressor p53 transcriptional activity and can supress heat shock and glucocorticoid signalling pathways. Additionally offers protection to cells from p53 mediated apoptosis. Active in vivo; protects mice from chemotherapy side effects associated with p53 induction. Also aryl hydrocarbon receptor (AHR) agonist.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide
  • CAS Nummer:63208-82-2
  • Molekulargewicht:367,3 g/mol
Spezifikationen für diese Produktfamilie

Spezifikation Testergebnisse

  • Constituents::
    100% Pifithrin-alpha-HBr
Product Family Options

Produktinformationen

  • Verfügbarkeit
    Preis
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 10 mg
    1 * 10 mg - 180,00 €
    180,00 €
    1 * 10 mg
       180,00 €

    Spezifikationen:
    Marketsource
    Marketsource
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 50 mg
    1 * 50 mg - 682,00 €
    682,00 €
    1 * 50 mg
       682,00 €

    Spezifikationen:
    Marketsource
    Marketsource