Express Eingabe
Austria
AuftragsverfolgungKontakt
Bromothymol blue pH-Indikator

Bromothymol blue pH-Indikator

Anbieter:   Abcam
Bromothymol blue pH-Indikator
Anbieter:  Abcam
Zertifikate

2 Optionen unten verfügbar

Product Details & Documents

pH indicator for weak acids and bases (pH range from 6,0 (yellow) to 7,6 (blue)). Also used as a stain to define cell walls or nuclei under the microscope.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Nummer:76-59-5
  • Molekulargewicht:624,38 g/mol
Spezifikationen für diese Produktfamilie

Spezifikation Testergebnisse

  • Constituents::
    100% Bromothymol blue free acid
Product Family Options

Produktinformationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.