Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Lomeguatrib ≥98% MGMT inhibitor

Anbieter:   Abcam
Lomeguatrib ≥98% MGMT inhibitor
Anbieter:  Abcam
Zertifikate

2 Optionen unten verfügbar

Product Details & Documents

Potent O⁶-methylguanine-DNA methyltransferase (MGMT) inhibitor (IC₅₀ =9 nM). Inhibits recombinant human ATase (IC₅₀ =5 nM). Shows antitumor effects in vivo. Enhances antitumor effects of temozolomide.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  6-((4-Bromothiophen-2-yl)methoxy)-9H-purin-2-amine
  • CAS Nummer:192441-08-0
  • Molekulargewicht:326,17 g/mol
Spezifikationen für diese Produktfamilie

Spezifikation Testergebnisse

  • Constituents::
    100% 6-[(4-bromothiophen-2-yl)methoxy]-7H-purin-2-amine
Product Family Options

unology General

Produktinformationen

  • Verfügbarkeit
    Preis
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 5 mg
    1 * 5 mg - 108,00 €
    108,00 €
    1 * 5 mg
       108,00 €

Produktinformationen

  • Verfügbarkeit
    Preis
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 25 mg
    1 * 25 mg - 410,00 €
    410,00 €
    1 * 25 mg
       410,00 €