Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Polymyxin B sulphate cationic surfactant antibiotic agent

Marketsource
Anbieter:   Abcam
Polymyxin B sulphate cationic surfactant antibiotic agent
Anbieter:  Abcam
Marketsource
Zertifikate

3 Optionen unten verfügbar

Product Details & Documents

effects against Gram-negative bacteria in vivo.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Nummer:1405-20-5
  • Molekulargewicht:1385,61 g/mol
Spezifikationen für diese Produktfamilie

Spezifikation Testergebnisse

  • Constituents::
    100% Polymyxin B sulfate
Product Family Options

unology General

Produktinformationen

  • Verfügbarkeit
    Preis
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 250 mg
    1 * 250 mg - 103,00 €
    103,00 €
    1 * 250 mg
       103,00 €

    Spezifikationen:
    Marketsource
    Marketsource
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 g
    1 * 1 g - 205,00 €
    205,00 €
    1 * 1 g
       205,00 €

    Spezifikationen:
    Marketsource
    Marketsource
  • Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 5 g
    1 * 5 g - 892,00 €
    892,00 €
    1 * 5 g
       892,00 €

    Spezifikationen:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.