Express Eingabe
Austria
AuftragsverfolgungKontakt
4'-Hydroxy-3'-methoxyacetophenone ≥99% NADPH-oxidase inhibitor

4'-Hydroxy-3'-methoxyacetophenone ≥99% NADPH-oxidase inhibitor

Artikelnr. ABCAAB120615-1
Marketsource
Anbieter:   Abcam
4'-Hydroxy-3'-methoxyacetophenone ≥99% NADPH-oxidase inhibitor
Artikelnr. ABCAAB120615-1
Anbieter:  Abcam
Marketsource
CAS Nummer:  498-02-2
4'-Hydroxy-3'-methoxyacetophenone ≥99% NADPH-oxidase inhibitor
CAS Nummer:  498-02-2
Preis
66,70 €
Einloggen für Verfügbarkeit
1 * 1 g
Product Details & Documents

Selective NADPH-oxidase inhibitor (IC₅₀ =10 μM). Inhibits production of reactive oxygen species. Also elicits a range of in vitroand in vivoanti-inflammatory effects.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyme:  4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
  • CAS Nummer:498-02-2
  • Molekulargewicht:166,17 g/mol
Spezifikationen

Spezifikation Testergebnisse

  • Constituents::
    100% Apocynin
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.