Express Eingabe
Austria
AuftragsverfolgungKontakt
Nuclear Extraction Kit, Abcam | Avantor

Nuclear Extraction Kit, Abcam

Nuclear Extraction Kit
Artikelnr. ABCAAB221978-100
Marketsource
Anbieter:   Abcam
Nuclear Extraction Kit, Abcam
Artikelnr. ABCAAB221978-100
Anbieter:  Abcam
Marketsource
CAS Nummer:  
Nuclear Extraction Kit, Abcam

Nuclear Extraction Kit

CAS Nummer:  
Preis
211,00 €
Einloggen für Verfügbarkeit
1 * 100 Tests
Product Details & Documents

This Nuclear Extraction Kit is formulated for the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates that can be used successfully in our transcription factor assay kits.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spezifikationen

Spezifikationen

  • Bezeichnung:
    Nuclear Extraction Kit
  • Cat. No.:
    ABCAAB221978-100
  • Lagertemperatur:
    Store at Multi.

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.