AuftragsverfolgungKontakt
Flow Through Cell | Avantor
Flow Through Cell
Flow through cell, glass body
Artikelnr. SENK255-75
New Product
Anbieter:   SENTEK
Flow Through Cell
Artikelnr. SENK255-75
Anbieter:  SENTEK
New Product
Flow Through Cell
Flow through cell, glass body
Preis
108,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Glass flow through cell for 12 mm pH and conductivity electrodes.
Designed for general purpose applications, particularly good for measuring conductivity in pure water.
Achtung! Bitte beachten: Not for pressure applications - only use to drain.
Spezifikationen
  • Bezeichnung:
    Flow through cell, glass body