ContactUsLinkComponent
SureSTART™ Screw Cap, 24 mm, Level 2 High-throughput Applications
Catalog # HYPE6ASC24-CTPAL
New Product
SureSTART™ Screw Cap, 24 mm, Level 2 High-throughput Applications
Catalog # HYPE6ASC24-CTPAL
New Product
Specifications
  • Bezeichnung:
    Level 2 Screw caps for high-throughput applications
  • Material:
    PP
  • Verschluss-Farbe:
    Weiß
  • VE:
    100
Specifications
About this item
SureSTART vials, caps, inserts, septa, and kits are available in three performance levels. Now you can choose the products that best suit your analytical, performance, or budget needs.
Use Level 2 Thermo Scientific™ SureSTART™ 24 mm screw caps with screw vials that have a 24 mm opening for your high-throughput applications, when robustness and reproducibility are key.
SureSTART Level two vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.