Express Eingabe
Austria
AuftragsverfolgungKontakt
Human Native Collagen VI | Avantor

Human Native Collagen VI

Artikelnr. ABCAAB7538-500
Marketsource
Anbieter:   Abcam
Human Native Collagen VI
Artikelnr. ABCAAB7538-500
Anbieter:  Abcam
Marketsource
CAS Nummer:  
Human Native Collagen VI
Human Native Collagen VI
CAS Nummer:  
Preis
574,00 €
Einloggen für Verfügbarkeit
1 * 500 µG
Product Details & Documents

Native Human Collagen VI protein is a Human Full Length protein with >95% and suitable for ELISA, IP, IHC, SDS-PAGE and western blot. Save time and ensure accurate results - use our Collagen VI (COL6A1-COL6A3) protein as a control -Does not cross react with other collagens.

Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Spezifikationen

Spezifikationen

  • Konjugation:
    Unconjugated
  • Form:
    Liquid
  • Transporttemperatur:
    20
  • Validierte Anwendungen:
    ELISA, IHC-P, IP, SDS-PAGE, WB
  • Spezies:
    Human
  • Konzentration:
    1 mg/ml
  • Zusammensetzung:
    Preservative: 0,01% Sodium azide, Constituents: 3% Acetic acid
  • Cat. No.:
    ABCAAB7538-500
  • Tierfrei:
    N
  • Protein/Peptid-Typ:
    Nativ
  • Lagerbedingungen:
    Store at 4 °C
  • Protein/Peptid-Name:
    Collagen VI
  • Proteinsynonym:
    Collagen alpha-1(VI) chain|COL6A1

Spezifikationen