Express Eingabe
Austria
AuftragsverfolgungKontakt
Human Recombinant PF4 (from E. coli)

Human Recombinant PF4 (from E. coli)

Catalog # ABCAAB9562-20
Marketsource
Supplier:   Abcam
Human Recombinant PF4 (from E. coli)
Catalog # ABCAAB9562-20
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant PF4 (from E. coli)
Human Recombinant PF4 (from E. coli)
CAS Number:  
Price
405,00 €
Log in for availability
1 * 20 µG
Product Details & Documents

Recombinant human PF4 protein is a Human Full Length protein, in the 31 to 101 aa range, expressed in Escherichia coli, with >98% purity and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Cat. No.:
    ABCAAB9562-20
  • Ursprung:
    E. coli
  • Tierfrei:
    N
  • Konjugation:
    Unconjugated
  • Validierte Anwendungen:
    FuncS, SDS-PAGE
  • Purity:
    >98%
  • Spezies:
    Human
  • Protein/Peptid-Typ:
    Rekombinant
  • Lagerbedingungen:
    –80 °C
  • Protein/Peptid-Name:
    PF4
  • Biologische Aktivität:
    Standard
  • Proteinsynonym:
    CXCL4|SCYB4|Platelet factor 4|PF-4|C-X-C motif chemokine 4|Iroplact|Oncostatin-A

Specifications