Express Eingabe
Austria
AuftragsverfolgungKontakt
Lincomycin ELISA Kit | Avantor

Lincomycin ELISA Kit

Lincomycin ELISA kit
Artikelnr. ANTIA327221-96
New Product
Anbieter:   Antibodies.com
Lincomycin ELISA Kit
Artikelnr. ANTIA327221-96
Anbieter:  Antibodies.com
New Product
CAS Nummer:  
Lincomycin ELISA Kit

Lincomycin ELISA kit

CAS Nummer:  
Preis
350,00 €
Einloggen für Verfügbarkeit
1 * 96 Tests
Product Details & Documents

Lincomycin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Lincomycin in egg, honey, feed, tissue, milk, milk powder, serum, urine and various sample types.

Lincomycin ELISA Test Kit employs the competitive enzyme immunoassay technique for the quantitative measurement of Lincomycin in egg, honey, feed, tissue, milk, milk powder, serum, urine and various sample types. The 96-well microtiter plate has been pre-coated with Lincomycin. During the incubation, Lincomycin present in the samples or standards competes with the fixed amount of immobilised Lincomycin for binding sites of the Anti-Lincomycin Antibody, a HRP-Conjugated Secondary Antibody is added to the wells at the same time. The more Lincomycin present in a sample or standard, the less Anti-Lincomycin Antibody that binds to the plate. Following incubation, unbound HRP-Conjugated Secondary Antibody is removed by washing. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Lincomycin present in each sample or standard. The concentration of Lincomycin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Achtung! Bitte beachten: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Spezifikationen

Spezifikationen

  • Bezeichnung:
    Lincomycin ELISA kit
  • Reaktivität der primären Antikörper:
    Other
  • Shelf life:
    Store for 6 months at +4 °C.
  • Testdauer:
    Multiple steps
  • Time to Results:
    10 - 30 min
  • Cat. No.:
    ANTIA327221-96
  • Lagertemperatur:
    +4 °C
  • Format:
    Pre-coated
  • Target protein:
    Lincomycin
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Competitive
  • Nachweisgrenze:
    0,2 ppb
  • Probentyp:
    Egg, honey, feed, tissue, milk, milk powder, serum, urine and various sample types.
  • Regulatory status:
    RUO

Spezifikationen