Mouse Procollagen III N-terminal propeptide (PIIINP) Antibody Pair - BSA and Azide free
ABCAAB253836-10X96
: Abcam
:
- Bezeichnung:Mouse Procollagen III N-terminal propeptide (PIIINP) antibody pair - BSA and Azide free
- Tests pro Kit:10×96 tests
Mouse Procollagen III N-terminal propeptide (PIIINP) Antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse Procollagen III N-terminal propeptide (PIIINP).