ContactUsLinkComponent
Cell culture dishes for adherent cells, Sarstedt SUREGrip design
Catalog # SARS83.3902
Supplier:  SARSTEDT
Cell culture dishes for adherent cells, Sarstedt SUREGrip design
Catalog # SARS83.3902
Supplier:  SARSTEDT
Specifications
  • Ø:
    100 mm
  • Wachstumsfläche:
    58 cm²
  • Verpackt:
    20/Beutel
  • Bezeichnung:
    Standard-Gewebekulturschale, ohne Gitter, SUREGrip Design, rot
  • Höhe:
    20 mm
  • VE:
    300
Specifications
About this item
PS. Standard TC treated dishes with convenient handling due to a raised ring around the circumference of the dish base, enabling an easy and secure grip of both the dish and the lid, even from a whole stack.