Calibration weight sets, OIML F1, Premium line
Supplier: Mettler - Toledo
Certificates
About this item
These weight sets made of premium, vacuum melted stainless steel ensures an anti-corrosive surface with low magnetisation and susceptibility values.
- Density of 8,0 kg/dm3
- Magnetic susceptibility of less than 0,01
- Ideal for balance calibration and testing
The proven one-piece design (monobloc) guarantees best long-term stability and accurate testing results.
Frequently Bought Together