Filter papers for fat analysis
Supplier: MACHEREY-NAGEL
Certificates
About this item
Smooth folded filter papers for fat analysis.
MN 615 ff paper is particularly suited for the analysis of fats. A special treatment with organic solvents guarantees that these filters are practically free of fats and resins (ether-soluble residue < 0,1 mg for a 270 mm filter circle).
MN 715 paper is also suited for the analysis of fats. Careful selection of the raw materials ensures a low ether-soluble residue for these filters.
Specifications
- Catalog No:
- MANA528027
- MANA528007
- MANA591015
- MANA528009
- MANA528005
- MANA528015
- MANA591027
- MANA528018
- MANA561032
- MANA591018
- MANA528011
- MANA528012
- MANA591024
- MANA591007
- MANA591009
- MANA528024
- MANA591005
- MANA591011
- MANA591012
- Gewicht:
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- Stärke:
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- Filtriergeschwindigkeit:
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s
- 22 s