Manual syphon pump
Lieferant: NEW PIG
Zertifikate
Über diesen Artikel
Save time and reduce waste with this chemical-resistant syphon pump.
- Durable polyethylene construction resists chemicals to maximise life of pump
- For water based liquids and general-purpose chemicals
- 380 mm long suction tube is ideal for smaller containers or drums up to 25 litres
- 534 mm delivery hose for accurate dispensing
- Easy-to-use manual suction operation; an economical and light-weight solution for every budget
It delivers mild solvents, light oils, corrosives, dilute acid/caustics, liquid waxes and disinfectants at up to 20 L/min.
Frequently Bought Together