Cotton dispensing jars, with lid
Catalog # HECH44295010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Ext.Ø:100 mm
- Höhe:100 mm
- VE:1
Specifications
About this item
Jars for cotton dispensing, made of glass with stainless steel lid.
- Sterilisable (without cotton)
- Good chemical resistance