Urine collection container
with handle and lid, 2000 ml
Artikelnr. HECH44116010
Anbieter: GLASWARENFABRIK KARL HECHT
Springe zu:
Product Details & Documents
PP, transparent, with white cover made of polycarbonate.
- Good chemical resistance
- Good temperature resiatnce
Urine collecting container, PP, with handle and embossed scale.
Spezifikationen
- Kapazität:2000 ml
- Bezeichnung:with handle and lid